Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RA Mouse anti-Rat, BV480, Clone: OX-33, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 746711
Description
CD45RA Monoclonal antibody specifically detects CD45RA in Rat samples. It is validated for Flow Cytometry.
Specifications
| CD45RA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Rat | |
| Purified |
| Flow Cytometry | |
| OX-33 | |
| Brilliant Violet 480 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction