Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD45RB Ab-3, Mouse Monoclonal Antibody

Catalog No. MS1846R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1846R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1846R1 Supplier Epredia™ Supplier No. MS1846R1

Target human CD45RB in immunohistochemistry applications with Epredia™ CD45RB Ab-3, Mouse Monoclonal Antibody.

CD45RB is expressed on mature B lymphocytes and the majority of lymphomas and leukemias of B-cell origin. This antigen is also expressed on the surface of cells of myeloid lineage and in the cytoplasm of tissue macrophages.

Host Species: Mouse

Clone: PD7/26

Isotype: IgG1/κ

Species Reactivity: Human. Others not known

Epitope: Exon B

Immunogen: Human peripheral blood lymphocytes maintained in T-cell growth factor

Molecular Weight: 170 kDa

Positive Control: Tonsil or lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD45RB Ab-3
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PD7/26
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Human peripheral blood lymphocytes maintained in T cell growth factor
Quantity 1 mL
Regulatory Status IVD
Research Discipline Hematopoietic
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.