Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RB Antibody (rPTPRC/1132), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308909MFV500
Description
CD45RB Monoclonal antibody specifically detects CD45RB in Human samples. It is validated for Flow CytometrySpecifications
CD45RB | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
rPTPRC/1132 | |
Flow Cytometry | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Recombinant full-length human CD45RB protein (Uniprot: P08575) | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction