Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD45RB Monoclonal Antibody (MEM-55), PE
GREENER_CHOICE

Catalog No. 01676364
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-364 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-364 Supplier Invitrogen™ Supplier No. MA119637
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes a siliadase-sensitive extracellular epitope of CD45RB, a 180-240 kDa single chain type I membrane glycoprotein, variant of CD45 (CD45RB isoform). CD45RB is expressed on a subset of T lymphocytes, B lymphocytes, monocytes, macrophages, granulocytes and dendritic cells.

CD45RB is a receptor-type protein tyrosine phosphatase, CD45 glycoprotein. CD45 is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases, promotes cell survival by modulating integrin-mediated signal transduction pathway and is also involved in DNA fragmentation during apoptosis. CD45 isoforms differ in their extracellular domains, but share identical transmembrane and cytoplasmic domains. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45RB is expressed in microglia and inflammatory cells. In addition to involvement in immunoreceptor signaling, CD45 is important in promoting cell survival by modulating integrin-mediated signal transduction pathway and is involved in DNA fragmentation during apoptosis. CD45 is expressed on cells of the human hematopoietic lineage with the exception of mature red cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RB
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-55
Concentration mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene Ptprc
Gene Accession No. P08575
Gene Alias B220; CD45; CD45 antigen; CD45R; GP180; LCA; L-CA; Leukocyte common antigen; loc; LY5; Ly-5; Lymphocyte antigen 5; lymphocyte common antigen; Lyt-4; membrane tyrosine phosphatase; protein tyrosine phosphatase receptor type C; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T200; T200 glycoprotein; T200 leukocyte common antigen
Gene Symbols Ptprc
Host Species Mouse
Immunogen Human thymocytes and T lymphocytes.
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5788
Target Species Human, Monkey
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.