Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RC Antibody (GL24), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP127991
Description
CD45RC Monoclonal specifically detects CD45RC in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.Specifications
CD45RC | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500 | |
PTPRC | |
Activated DBA/2 B cells | |
0.25 mg | |
Mesenchymal Stem Cell Markers, Stem Cell Markers | |
5788 | |
Mouse | |
Purified |
Flow Cytometry, Immunoprecipitation | |
GL24 | |
Biotin | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Rat | |
Multi-step | |
RUO | |
Primary | |
An exon C-dependent epitope of CD45 glycoprotein | |
Store at 4C. Do not freeze. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction