Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RC Antibody (OX22), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1206406UV
Description
CD45RC Monoclonal specifically detects CD45RC in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
CD45RC | |
Monoclonal | |
DyLight 350 | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB120-6406 recognizes rat CD45RC, the high molecular weight form of the leucocyte common antigen. This antigen is found on B cells, approximately 50% of bone marrow cells, all CD8+ve T cells, but splits CD4+ve T cells into two populations,CD4+ CD45RC (Th 1-like) and CD4+ CD45RC low (Th-2 like). This product is routinely tested in flow cytometry on rat splenocytes. | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
OX22 | |
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
PTPRC | |
Rat lymphocytes stimulated in culture with phytohaemagglutinin. | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788 | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction