Learn More
CD45RC Mouse Anti-Rat, Brilliant Violet 605, Clone: OX-22, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 742488
Description
The OX-22 antibody reacts with CD45RC on pre-B lymphocytes, B cells, CD8+ T suppressor/cytotoxic cells, and a subset of CD4+ T helper (Th) lymphocytes. It weakly reacts with thymocytes. CD45RC is a high-molecular-weight isoform of CD45 (Leukocyte Common Antigen); its level of expression distinguishes subpopulations of CD4+ T cells with Th1-like and Th2-like effector functions. Levels of expression of CD45RC have also been reported to distinguish resting from activated T cells at various stages of maturation. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications
CD45RC | |
OX-22 | |
Brilliant Violet 605 | |
CD45RC | |
PHA-activated rat lymph node cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.