Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RO Antibody (UCHL-1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP233104R
Description
CD45RO Monoclonal specifically detects CD45RO in Human, Chimpanzee, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CD45RO | |
Monoclonal | |
DyLight 550 | |
P08575 | |
PTPRC | |
Interleukin-2-dependent human T lymphocytes were used as the immunogen for the CD45RO antibody. | |
Protein A or G purified | |
RUO | |
Primary | |
Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO) (4th Leucocyte Typing Workshop: Code No. N31). The epitope recognized by this antibody is sensitive to neuraminidase digestion. This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. Reportedly, it is useful to identify T-cell lymphomas and leukemias. It rarely stains NK cells or B-cell lymphomas. | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
UCHL-1 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1-5 μg/mL for coating, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Mouse | |
182 kDa | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Microglia Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
5788 | |
Human, Primate | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction