Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD45RO Mouse anti-Human, BV750, Clone: UCHL1, BD Biosciences™

Catalog No. BDB746942 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB746942 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB746942 Supplier BD Biosciences Supplier No. 746942
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45RO
Applications Flow Cytometry
Classification Monoclonal
Clone UCHL1
Conjugate Brilliant Violet 750
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD45R; PTPRC; LCA; Leukocyte common antigen; GP180; LY5; T200
Host Species Mouse
Immunogen Human IL-2-dependent T-cell line
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.