Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RO Mouse anti-Human, Clone: UCHL-1+T200/797, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14693
Description
Mouse monoclonal antibody raised against human CD45RO.
- High specificity
- High reactivity
Specifications
CD45RO | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
B220/CD45/CD45R/GP180/LCA/LY5/T200 | |
Mouse | |
Protein A/G purification | |
RUO | |
5788 | |
Human | |
Liquid |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
UCHL-1+T200/797 | |
Mouse monoclonal antibody raised against human CD45RO. | |
In 10 mM PBS (0.05% BSA and 0.05% azide). | |
PTPRC | |
Cultured human T-cells from an IL-2-dependent T-cell line (CA1) (UCHL-1) and Human T-lymphocytes (T200/797). | |
100 μg | |
Primary | |
Recognizes a 180-185kDa protein, identified as isoform of leukocyte common antigen (CD45RO). This antibody reacts with mature activated T-cells, most thymocytes, and a sub-population of resting T-cells within both CD4 and CD8 subsets. It shows no reactivity with normal B or natural killer cells, but reacts with granulocytes and monocytes. It rarely stains NK cells or B-cell lymphomas. | |
Store at 4°C. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction