Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45RO Mouse anti-Human, RB545, Clone: UCHL1, BD Horizon™
Mouse Monoclonal Antibody
$396.00
Specifications
Antigen | CD45RO |
---|---|
Clone | UCHL1 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB545 |
Description
CD45RO Monoclonal antibody specifically detects CD45RO in Human samples. It is validated for Flow cytometrySpecifications
CD45RO | |
Flow Cytometry | |
RB545 | |
Mouse | |
Human | |
CD45R; PTPRC; LCA; Leukocyte common antigen; GP180; LY5; T200 | |
Human IL-2-dependent T-cell line | |
Primary | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
UCHL1 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
5788 | |
IgG2a κ | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title