Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD46 Antibody (MCP/931), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311539F
Description
CD46 Monoclonal antibody specifically detects CD46 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
CD46 | |
Monoclonal | |
FITC | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
MCP/931 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen | |
Recombinant human CD46 protein (Uniprot: P15529) | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Immunology | |
4179 | |
Store at 4°C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction