Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD46 Antibody (MM.1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311191
Description
CD46 Monoclonal specifically detects CD46 in Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.Specifications
CD46 | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay | |
AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen | |
Second and third SCR domains of MCP fused to IgG-Fc; MCP23-Ig | |
50 μg | |
Cancer, Cell Biology, Cellular Markers, Immunology | |
4179 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Western Blot, Immunofluorescence, Immunohistochemistry (Frozen), Immunoassay | |
MM.1 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction