Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD46 Monoclonal Antibody (MEM-258), Biotin
GREENER_CHOICE

Catalog No. 01676254
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-254 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-254 Supplier Invitrogen™ Supplier No. MA119520
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an extracellular epitope on SCR4 (the membrane-proximal SCR) domain of CD46 (Membrane cofactor protein). CD46 is 56-66 kDa dimeric transmembrane protein expressed on T and B lymphocytes, platelets, monocytes, granulocytes, endothelial cells, epithelial cells and fibroblast; it is negative on erythrocytes.

CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD46
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MEM-258
Concentration 1 mg/mL
Conjugate Biotin
Formulation PBS with 15mM sodium azide; pH 7.4
Gene CD46
Gene Accession No. P15529, Q6VE48
Gene Alias AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antigen
Gene Symbols CD46
Host Species Mouse
Immunogen T cell line HPB-ALL.
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 280851, 4179
Target Species Human, Bovine
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.