Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD46 Monoclonal Antibody (MEM-258), PE
GREENER_CHOICE

Catalog No. PIMA182564
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA182564 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA182564 Supplier Invitrogen™ Supplier No. MA182564
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD46 antibody, clone MEM-258 recognizes the human CD46 cell surface antigen, a broadly expressed glycoprotein also known as membrane co-factor protein (MCP).

CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD46
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-258
Concentration Conc. Not Determined
Conjugate PE
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene CD46
Gene Accession No. P15529
Gene Alias AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antigen
Gene Symbols CD46
Host Species Mouse
Immunogen HPB-ALL cell line.
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4179
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.