Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD46, Mouse anti-Human, Clone: TRA-2-10, Millipore Sigma™

Catalog No. MABF218225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF218225 25 μL
MABF2182100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF218225 Supplier MilliporeSigma Supplier No. MABF218225UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD46, clone TRA-2-10, Cat. No. MABF2182, is a mouse monoclonal antibody that detects Membrane cofactor protein (CD46) and has been for use in Flow Cytometry, Immunoprecipitation, Immunohistochemistry, and Inhibition assay.

Membrane cofactor protein (UniProt: P15529; also known as TLX, Trophoblast leukocyte common antigen, CD46) is encoded by the CD46 (also known as MCP; MIC10) gene (Gene ID: 4179) in human. CD46 is a single-pass type I membrane protein that is expressed by all cells except erythrocytes. It acts as a cofactor for complement factor I, a serine protease that protects autologous cells against complement-mediated injury by cleaving C3b and C4b deposited on host tissue. It also acts as a costimulatory factor for T-cells which induces the differentiation of CD4+ into T-regulatory 1 cells. CD46 is synthesized with a signal peptide (aa 1-34), which is cleaved off in the mature form. The mature form contains an extracellular domain (aa 35-343), a transmembrane domain (aa 344-366), and a cytoplasmic domain (aa 367-392). It also contains four Sushi domains where Sushi domains 1 and 2 are required for interaction with human adenovirus B PIV/FIBER protein and with Measles Virus H protein, domains 2 and 3 are essential for Herpes virus 6 binding, and domains 3 and 4 are required for interaction with Streptococcus Pyrogens M protein and are the most important for interaction with C3b and C4b. CD46 can be glycosylated on Asn 83, 114, and 273 and N-glycosylation on Asn 114 and 273 are required for its cytoprotective function. N-glycosylation on Asn 114 is also required for Measles virus binding and N-glycosylation on Asn-273 is required for Neisseria binding. CD46 can also be extensively O-glycosylated in the Ser/Thr-rich domain, which is required for Neisseria binding, but not for Measles virus binding. Mutations in CD46 gene have been linked to hemolytic uremic syndrome atypical 2 that has been characterized by microangiopathic hemolytic anemia, thrombocytopenia, and renal failure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD46
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays
Classification Monoclonal
Clone TRA-2-10
Conjugate Unconjugated
Dilution Inhibition Analysis: A representative lot blocked Measles virus infection in BC1 cells. (Manchester, M., et. al. (1995). Proc Natl Acad Sci U S A. 92(6):2303-7). Immunohistochemistry Analysis: A representative lot detected CD46 in Immunohistochemistry applications (Kemper, C., et. al. (2001). Clin Exp Immunol. 124(2):180-9).Immunoprecipitation Analysis: A representative lot immunoprecipitated CD46 in Immunoprecipitation applications (Andrews, P.W., et. al. (1985). Ann Hum Genet. 49(Pt 1):31-9).Flow Cytometry Analysis: A representative lot detected CD46 in Flow Cytometry applications (Manchester, M., et. al. (1995). Proc Natl Acad Sci U S A. 92(6):2303-7).
Formulation Purified mouse monoclonal antibody IgG1 in PBS without preservatives.
Gene Alias Membrane cofactor protein;TLX;Trophoblast leukocyte common antigen
Gene Symbols CD46;MCP;MIC10
Host Species Mouse
Immunogen 2102Ep human embryonal carcinoma cells.
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_002380
Test Specificity Clone TRA-2-10 specifically detects CD46 in human cells.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.