Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578985
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578985 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578985 Supplier Invitrogen™ Supplier No. PA578985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human K562 whole cell, human A549 whole cell, human A431 whole cell. IHC: human placenta tissue, human prostatic cancer tissue. ICC/IF: T-47D cell .

CD46 (MCP, membrane cofactor protein) is a multifunctional cell surface transmembrane protein that binds and inactivates C3b and C4b complement fragments, regulates T cell-induced inflammatory responses by either inhibiting (CD46-1 isoform) or increasing (CD46-2 isoform) the contact hypersensitivity reaction. CD46 also serves as a receptor for several human pathogens (both bacteria and viruses), and its ligation alteres T lymphocyte polarization toward antigen-presenting cells or target cells, inhibiting lymphocyte function. CD46 is a protector of placental tissue and is also expressed on the inner acrosomal membrane of spermatozoa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD46
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.01mg sodium azide
Gene CD46
Gene Accession No. O88174, P15529, Q9Z0M4
Gene Alias AHUS2; antigen identified by monoclonal antibody TRA-2-10; brain microvascular membrane cofactor; brain microvascular membrane cofactor protein CD46; Cd46; CD46 antigen, complement regulatory protein; CD46 complement regulatory protein; CD46 molecule; CD46 molecule, complement regulatory protein; CD46 splice variant; complement membrane cofactor protein; complement regulatory protein; Mcp; MCP; complement activation regulator; measles virus receptor; membrane cofactor protein; membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen); membrane cofactor protein variant 10; membrane cofactor protein variant 7; membrane cofactor protein variant 8; membrane cofactor protein variant 9; MGC26544; MIC10; soluble membrane cofactor protein variant 11; soluble membrane cofactor protein variant 12; soluble membrane cofactor protein variant 13; TLX; TRA2.10; trophoblast leucocyte common antigen; trophoblast leukocyte common antigen; trophoblast-lymphocyte cross-reactive antigen
Gene Symbols CD46
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the C-terminus of human CD46 (366-392aa YRYLQRRKKKGTYLTDETHREVKFTSL).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17221, 29333, 4179
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.