Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD47 Antibody (B6H12.2) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 3 publications
$281.00 - $562.50
Specifications
Antigen | CD47 |
---|---|
Clone | B6H12.2 |
Concentration | 1.0 mg/mL |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23110625
![]() |
Novus Biologicals
NBP23110625UG |
25 μg |
Each for $281.00
|
|
|||||
NBP231106
![]() |
Novus Biologicals
NBP231106 |
0.1 mg |
Each for $562.50
|
|
|||||
Description
CD47 Monoclonal specifically detects CD47 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Flow (Cell Surface), Immunocytochemistry, CyTOF-ready.Specifications
CD47 | |
1.0 mg/mL | |
Unconjugated | |
Mouse | |
Immunology, Signal Transduction | |
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
CD47 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
B6H12.2 | |
Monoclonal | |
Purified | |
RUO | |
Q08722 | |
961 | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title