Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD47 Antibody (CD47/6362R), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308218H
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
CD47 | |
Monoclonal | |
HRP | |
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
A recombinant fragment (around aa 18-135) of human CD47 protein (exact sequence is proprietary) (Uniprot: Q08722) | |
100 μg | |
Immunology, Signal Transduction | |
961 | |
Store at 4°C in the dark. | |
IgG κ |
ELISA, Immunohistochemistry (Paraffin) | |
CD47/6362R | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction