Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD47 Antibody [DyLight 594], Novus Biologicals™

Sheep Polyclonal Antibody
Supplier: Novus Biologicals AF4670M
Description
CD47 Polyclonal antibody specifically detects CD47 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Neutralization,CyTOF,Gene Knock-OutSpecifications
CD47 | |
Polyclonal | |
50mM Sodium Borate | |
Sheep | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Western Blot, Neutralization, CyTOF, Gene Knock-Out | |
DyLight 594 | |
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
Mouse myeloma cell line NS0-derived recombinant human CD47, Gln19-Pro139, Accession # Q08722 | |
0.1 mL | |
Immunology, Signal Transduction | |
961 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction