Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD47 Antibody (TH17A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260921
Description
CD47 Monoclonal specifically detects CD47 in Porcine, Bovine, Goat, Llama, Bison, Alpaca, Water Buffalo, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD47 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
Mouse | |
Unpurified | |
Immunology, Signal Transduction | |
961 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
TH17A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
CD47 | |
Human mononuclear leukocytes | |
0.1 mg | |
Primary | |
Pig, Bovine, Goat, Llama, Bison, Alpaca, Water Buffalo, Sheep | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction