Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD47 Monoclonal Antibody (2D3), NovaFluor™ Yellow 755, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$194.00 - $430.00
Specifications
| Antigen | CD47 |
|---|---|
| Clone | 2D3 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
H065T03Y08A
|
Invitrogen™
H065T03Y08A |
100 Tests |
Each for $430.00
|
|
|||||
H065T02Y08A
|
Invitrogen™
H065T02Y08A |
25 Tests |
Each for $194.00
|
|
|||||
Description
CD47 Monoclonal antibody specifically detects CD47 in Human samples. It is validated for Flow CytometrySpecifications
| CD47 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q08722 | |
| 961 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| 2D3 | |
| Flow Cytometry | |
| NovaFluor Yellow 755 | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 9130415E20Rik; AA407862; AI848868; antigen identified by monoclonal antibody 1D8; antigenic surface determinant protein OA3; AW108519; B430305P08Rik; Cd47; CD47 antigen; CD47 antigen (Rh-related antigen, integrin-associated signal transducer); CD47 antigen/integrin-associated protein; CD47 glycoprotein; CD47 molecule; CD47/IAP; IAP; integrin associated protein; integrin-associated protein; integrin-associated signal transducer; Itgp; Leukocyte surface antigen CD47; MER6; OA3; Protein MER6; Rh-related antigen; sCD47; soluble CD 47; soluble CD47 | |
| IgG1 κ | |
| Affinity chromatography | |
| CD47 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title