Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Mouse anti-Human, Brilliant Violet 421, Clone: B6H12, BD

Catalog No. BDB566255 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566255 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566255 Supplier BD Biosciences Supplier No. 566255
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B6H12 monoclonal antibody specifically binds to CD47, a 42-52 kDa N-linked glycan protein. CD47 antigen, also known as integrin-associated protein (IAP), is expressed on all hematopoietic cells, including leukocytes, platelets and erythrocytes. It is also expressed on epithelial cells, endothelial cells, fibroblasts and many tumor cell lines. CD47 may play a role as a signal transducer in the regulation of cation fluxes across cell membranes and in the chemotactic and adhesive interactions of leukocytes with endothelial cells. The B6H12 antibody is capable of blocking phagocytosis of microparticles by peripheral blood granulocytes. It has also been reported to induce proliferation of CD3-activated T cells

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD47
Classification Monoclonal
Clone B6H12
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias IAP, CDw149, gp42, MER6, Neurophilin, OA3, Rh-related antigen
Host Species Mouse
Immunogen Purified human placental RGD-binding proteins
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.