Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD47 Mouse anti-Human, Clone: PF3.1, MilliporeSigma™

Catalog No. MABF958 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF958 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF958 Supplier MilliporeSigma Supplier No. MABF958
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-CD47, clone PF3.1 Antibody Immunoprecipitation, and Western Blotting for the detection of CD47.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD47
Applications Flow Cytometry
Classification Monoclonal
Clone PF3.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CD47,MER6
Host Species Mouse
Immunogen CD47 purified from human spleen cells (Liu, Y., et al. (2001). J. Biol. Chem. 276(43):40156-40166).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001768
Test Specificity Clone PF3.1 detects human CD47 by targeting CD47 extracellular domain. In contrast to clone C5/D5 (Cat. No. MABF959), clone PF3.1 is suitable for immunoblotting, but not suitable for neutralizing CD47-dependent cellular signaling, while both clones can stain cell surface CD47 (Liu, Y., et al. (2001). J. Biol. Chem. 276(43):40156-40166).
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.