Learn More
Invitrogen™ CD48 Monoclonal Antibody (HM48-1), APC-eFluor™ 780, eBioscience™, Invitrogen™
Description
Description: The HM48-1 monoclonal antibody reacts with the mouse CD48 antigen; also known as BCM1, Blast-1 (human), and OX-45 (rat). CD48, a member of the SLAM family and Ig superfamily, is a 45 kDa GPI-linked glycoprotein expressed on the majority of hematopoietic cells. Recent publications have reported differential expression of members of the SLAM family including CD48, CD150, and CD244 among functionally distinct bone marrow hematopoietic progenitors providing a useful tool for prediction of the primitiveness of hematopoietic progenitors based on the expression of these SLAM family members. Hematopoietic stem cells (HSC) are highly purified as CD150(+)CD244(-)CD48(-) cells while non-self-renewing multipotent hematopoietic progenitors (MPP) are CD244(+)CD150(-)CD48(-) and the most restricted progenitors are CD48(+)CD244(+)CD150(-). CD48 plays a critical role in adhesion and T cell activation. In the mouse, the primary counter-receptors for CD48 are CD2 and CD244. HM48-1 is reported to modulate in vitro and in vivo CD48 functions including blocking the CD48/CD2 and CD48/CD244 interactions, inhibiting the proliferative response of mitogen-activated spleen cells, providing a costimulation signal for T cells activated in vitro through their TCR, and prolonging cardiac allograft survival in vivo. Applications Reported: This HM48-1 antibody has been reported for use in flow cytometric analysis.
Specifications
Specifications
| Antigen | CD48 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HM48-1 |
| Concentration | 0.2 mg/mL |
| Conjugate | APC-eFluor 780 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Cd48 |
| Gene Accession No. | P18181 |
| Gene Alias | AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1 |
| Show More |
Frequently Asked Questions (FAQs)
Our options will depend on the samples you are analyzing.
If cell viability is not critical, you can store your stained samples at 4 degrees C or on ice overnight in the dark and analyze the following day.
For samples stained with eFluor organic fluorochromes, we recommend that cells be suspended in 100 uL of Flow Cytometry Staining Buffer (Cat. No. 00-4222) and 100 uL of eBioscience IC Fixation Buffer (Cat. No. 00-8222); samples can be incubated for up to 3 days at 4 degrees C in the dark. Alternatively, the 1-step Fix/Lyse Solution (Cat. No. 00-5333) can be used. This is a great option when working with whole blood but also works for other cell types.
The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.
The eFluor Organic Dyes (eFluor 450, APC-eFluor 780, PerCP-eFluor 710, eFluor 710) are conventional fluorochromes. In contrast, the eVolve line of products are Quantum dots.
As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.