Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD48 Monoclonal Antibody (HM48-1), NovaFluor™ Red 755, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
$194.00 - $375.00
Specifications
Antigen | CD48 |
---|---|
Clone | HM48-1 |
Concentration | 0.1 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
M031T03R06A
![]() |
Invitrogen™
M031T03R06A |
100 μg |
Each for $375.00
|
|
|||||
M031T02R06A
![]() |
Invitrogen™
M031T02R06A |
25 μg |
Each for $194.00
|
|
|||||
Description
CD48 Monoclonal antibody specifically detects CD48 in Mouse samples. It is validated for Flow CytometrySpecifications
CD48 | |
0.1 mg/mL | |
Monoclonal | |
Armenian Hamster | |
Mouse | |
P18181 | |
12506 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
HM48-1 | |
Flow Cytometry | |
NovaFluor Red 755 | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1 | |
IgG | |
Affinity chromatography | |
Cd48 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title