Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD48 Monoclonal Antibody (HM48-1), PE-eFluor™ 610, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 61048182
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-048-182 100 μg
61-048-180 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 61-048-182 Supplier Invitrogen™ Supplier No. 61048182
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

The HM48-1 MAb reacts with the mouse CD48 antigen; also known as BCM1, Blast-1 (human), and OX-45 (rat). CD48, a member of the SLAM family and Ig superfamily, is a 45 kDa GPI-linked glycoprotein expressed on the majority of hematopoietic cells. Recent publications have reported differential expression of members of the SLAM family including CD48, CD150, and CD244 among functionally distinct bone marrow hematopoietic progenitors providing a useful tool for prediction of the primitiveness of hematopoietic progenitors based on the expression of these SLAM family members. Hematopoietic stem cells (HSC) are highly purified as CD150(+)CD244(-)CD48(-) cells while non-self-renewing multipotent hematopoietic progenitors (MPP) are CD244(+)CD150(-)CD48(-) and the most restricted progenitors are CD48(+)CD244(+)CD150(-). CD48 plays a critical role in adhesion and T cell activation. In the mouse, the primary counter-receptors for CD48 are CD2 and CD244. HM48-1 is reported to modulate in vitro and in vivo CD48 functions including blocking the CD48/CD2 and CD48/CD244 interactions, inhibiting the proliferative response of mitogen-activated spleen cells, providing a costimulation signal for T cells activated in vitro through their TCR, and prolonging cardiac allograft survival in vivo.

CD48, also known as Blast-1, is a 45 kDa glycoprotein that is part of the SLAM and Ig superfamilies. It is a GPI-anchored protein broadly expressed on most hematopoietic cells, including lymphocytes and monocytes, but is weakly expressed on granulocytes and absent on platelets, fibroblasts, epithelium, and endothelium. CD48 plays a critical role in adhesion and T cell activation, serving as a high-affinity ligand for 2B4 and a low-affinity ligand for CD2. The interaction between 2B4 and CD48 among NK and NK-T cells regulates cell proliferation. CD48 is involved in eosinophil activation and its expression increases in several infectious diseases. It is also a marker for detecting defects in the GPI anchoring structure in patients with paroxysmal nocturnal hemoglobulinuria (PNH). Recent studies have highlighted the differential expression of SLAM family members, including CD48, CD150, and CD244, among bone marrow hematopoietic progenitors. This provides a useful tool for predicting the primitiveness of hematopoietic progenitors: hematopoietic stem cells (HSCs) are CD150(+)CD244(-)CD48(-), non-self-renewing multipotent progenitors (MPPs) are CD244(+)CD150(-)CD48(-), and the most restricted progenitors are CD48(+)CD244(+)CD150(-). Additionally, the 2B4-CD48 interaction may be involved in the pathology of allergic airway diseases, offering a potential pharmacological target for treating conditions like allergic rhinitis and asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD48
Applications Flow Cytometry
Classification Monoclonal
Clone HM48-1
Concentration 0.2 mg/mL
Conjugate PE-eFluor 610
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cd48
Gene Accession No. P18181
Gene Alias AI449234; AW610730; B-cell activation marker; BCM1; Bcm-1; BCM1 surface antigen; BLAST; BLAST1; BLAST-1; B-lymphocyte activation marker BLAST-1; Cd48; CD48 antigen; CD48 antigen (B cell membrane protein); CD48 antigen (B-cell membrane protein); Cd48 molecule; CD48d; hCD48; HM48-1; leukocyte antigen MEM-102; mCD48; MEM-102; MRC OX-45 surface antigen; sCD 48; sCD48; Sgp-60; Signaling lymphocytic activation molecule 2; Signaling lymphocytic activation molecule 2 (SLAMF2); SLAM family member 2; SLAMF2; soluble CD 48; soluble CD48; TCT.1
Gene Symbols Cd48
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12506
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.