Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD48 Mouse anti-Human, Clone: 156-4H9, Abnova™

Catalog No. 89035091 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-091 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-091 Supplier Abnova Corporation Supplier No. MAB15071
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against native human CD48.

  • High specificity
  • High reactivity

Specifications

Antigen CD48
Applications Flow Cytometry, Functional Assay, Immunofluorescence
Classification Monoclonal
Clone 156-4H9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native human CD48.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Functional Study Immunofluorescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Accession No. P09326
Gene Alias BCM1/BLAST/BLAST1/MEM-102/SLAMF2/hCD48/mCD48
Gene Symbols CD48
Host Species Mouse
Immunogen Native purified human CD48.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 962
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.