Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD48/SLAMF2 Antibody (156-4H9), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143403PEATT594
Description
CD48/SLAMF2 Monoclonal specifically detects CD48/SLAMF2 in Human samples. It is validated for Flow Cytometry.Specifications
CD48/SLAMF2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
CD48 | |
The immunogen for this antibody was CD48. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
156-4H9 | |
Flow Cytometry | |
BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
Mouse | |
Protein A or G purified | |
Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
962.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction