Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD48/SLAMF2 Antibody (331504), Allophycocyanin/Cy7, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3327APCCY7
Description
CD48/SLAMF2 Monoclonal antibody specifically detects CD48/SLAMF2 in Mouse samples. It is validated for Flow CytometrySpecifications
CD48/SLAMF2 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
331504 | |
Flow Cytometry | |
BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD48/SLAMF2, Phe23-Arg216, Accession # BAE96326 | |
0.1 mL | |
Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
962 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction