Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD48/SLAMF2 Antibody (7E1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | CD48/SLAMF2 |
---|---|
Clone | 7E1 |
Dilution | Western Blot 1:500-1:5000, ELISA |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
CD48/SLAMF2 Monoclonal antibody specifically detects CD48/SLAMF2 in Human samples. It is validated for Western Blot, ELISASpecifications
CD48/SLAMF2 | |
Western Blot 1:500-1:5000, ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2 | |
A synthesized peptide derived from Human CD48/SLAMF2 [UniProt P09326] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
7E1 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers, Immunology, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
962 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title