Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ CD48/SLAMF2 Antibody Pair [HRP]
SDP

Catalog No. NB010395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010395 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010395 Supplier Novus Biologicals™ Supplier No. NBP27967815PLATES
Add to Cart
Add to Cart

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD48/SLAMF2.

Specifications

Assay Range 9.38 pg/mL
Assay Sensitivity 9.38-600 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CD48/SLAMF2.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias BCM1 surface antigen, BCM1Leukocyte antigen MEM-102, BLAST, BLAST1TCT.1, B-lymphocyte activation marker BLAST-1, CD48 antigen, CD48 antigen (B-cell membrane protein), CD48 molecule, hCD48, mCD48, MEM-102, SLAMF2
Gene ID (Entrez) 962
Gene Symbol CD48
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 34 ng of recombinant Human CD48/SLAMF2.
  • Capture Antibody: 0.2 mg/mL of mouse anti-Human CD48/SLAMF2 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human CD48/SLAMF2 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Quantity 15 Plates
    Regulatory Status RUO
    Research Discipline Cell Biology, Cellular Markers, Immunology, Stem Cell Markers
    Storage Requirements Storage is content dependent.
    Target CD48/SLAMF2
    Technique 1. Capture Antibody: Dilute to a working concentration of 1 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.2 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.