Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49a Monoclonal Antibody (CL7207)
GREENER_CHOICE

Catalog No. PIMA535041
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535041 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535041 Supplier Invitrogen™ Supplier No. MA535041
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: RQISRSFFSG TQERKVQRN Highest antigen sequence indentity to the following orthologs: Rat - 95%, Mouse - 89%.

Integrin alpha 1 (ITGA1) chain associates with the beta 1 (ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49a
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL7207
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Itga1
Gene Accession No. P56199
Gene Alias CD49 antigen-like family member A; CD49a; E130012M19Rik; integrin alpha 1; integrin alpha-1; integrin subunit alpha 1; integrin, alpha 1; Itga1; Laminin and collagen receptor; very late activation protein 1; Vla1; VLA-1
Gene Symbols Itga1
Host Species Mouse
Immunogen Synthetic Peptide corresponding to Human CD49a.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3672
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.