Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49a Mouse anti-Human, Brilliant Violet 650, Clone: SR84, BD Optibuild™

Catalog No. BDB742360 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742360 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742360 Supplier BD Biosciences Supplier No. 742360
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SR84 monoclonal antibody specifically recognizes CD49a (CD49 antigen-like family member A). CD49a is a ñ200 kDa type I transmembrane molecule that is encoded by ITGA1 ( /Integrin subunit alpha 1). CD49a is also known as very late antigen 1αsubunit (VLA-1) of the integrin family of cell adhesion molecules. CD49a associates with the integrinβ1 subunit (CD29) to form theα1/β1 heterodimer (CD49a/CD29), and serves as receptor for collagen and laminin-1. It is expressed on activated T cells, monocytes, neuronal cells and smooth muscle cells. CD49a has been reported to play a role in cell attachment during the development of both the central and peripheral nervous systems.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49a
Classification Monoclonal
Clone SR84
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Integrin α1, Integrin alpha-1, ITGA1, ITA1, VLA-1, VLA1
Host Species Mouse
Immunogen P-Selectin IgG1 Fusion
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.