Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49a Polyclonal Antibody, APC
GREENER_CHOICE

Catalog No. PIPA546899
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546899 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546899 Supplier Invitrogen™ Supplier No. PA546899
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Integrin alpha 2, rhIntegrin alpha 11, recombinant mouse (rm) Integrin alpha 2, and rmIntegrin alpha 11 is observed.

Integrin alpha 1 (ITGA1) chain associates with the beta 1 (ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49a
Applications Flow Cytometry
Classification Polyclonal
Conjugate APC
Formulation Buffered saline solution with BSA and 0.09% sodium azide
Gene Itga1
Gene Accession No. P56199
Gene Alias CD49 antigen-like family member A; CD49a; E130012M19Rik; integrin alpha 1; integrin alpha-1; integrin subunit alpha 1; integrin, alpha 1; Itga1; Laminin and collagen receptor; very late activation protein 1; Vla1; VLA-1
Gene Symbols Itga1
Host Species Sheep
Immunogen CHO-derived recombinant human Integrin alpha 1/CD49a Phe29-Pro1141.
Purification Method Antigen affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3672
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.