Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD49a Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100839
Description
Antibody detects endogenous levels of total ITGA1.
Integrin alpha 1 (ITGA1) chain associates with the beta 1 (ITGB1) chain to form a heterodimer that functions as a dual laminin/collagen receptor in neural cells and hematopoietic cells. ITGA1 has a 206-amino acid I domain in its N-terminal half, followed by 3 divalent cation-binding sites and a C-terminal transmembrane domain with a short cytoplasmic tail. It also has 28 potential N-glycosylation sites. Human ITGA1 was expressed in a mouse fibroblast cell line as a 180-kD protein. ITGA1 is involved in the early remodeling of osteoarthritic cartilage and plays an essential role in the regulation of mesenchymal stem cell proliferation and cartilage production. It also plays an essential role in the regulation of MSC proliferation and cartilage production.
Specifications
CD49a | |
Polyclonal | |
Unconjugated | |
Itga1 | |
CD49 antigen-like family member A; CD49a; E130012M19Rik; integrin alpha 1; integrin alpha-1; integrin subunit alpha 1; integrin, alpha 1; Itga1; Laminin and collagen receptor; very late activation protein 1; Vla1; VLA-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
109700, 25118, 3672 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P18614, P56199, Q3V3R4 | |
Itga1 | |
A synthesized peptide derived from human ITGA1(Accession P56199), corresponding to amino acid residues K766-S816. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction