Learn More
Invitrogen™ CD49b (Integrin alpha 2) Monoclonal Antibody (P1H5), PE, eBioscience™
Mouse Monoclonal Antibody
$370.00
Specifications
Antigen | CD49b (Integrin alpha 2) |
---|---|
Clone | P1H5 |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
12-050-042
![]() |
Invitrogen™
12050042 |
100 Tests |
Each of 1 for $370.00
|
![]() |
|||||
Description
Description: This P1H5 monoclonal antibody reacts with human CD49b, which is also known as integrin alpha 2. Integrins constitute a family of cell surface receptors that mediate adhesion between cells or between a cell and the extracellular matrix. These proteins are heterodimers consisting of two distinct subunits, alpha and beta. Integrins are expressed on hematopoietic, non-hematopoietic, and cancer cells. Integrin-mediated adhesion is regulated by changes in ligand affinity (or activation), cell shape, and integrin clustering and diffusion. Integrin alpha 2 forms a complex with the common beta 1 subunit to form the alpha2beta1 integrin, which binds type I collagen and laminin. Moreover, studies suggest that this complex suppresses metastasis in cancer and participates in T cell costimulation by activating the MAPK and PI3K pathways. The P1H5 monoclonal antibody has been reported to inhibit adhesion of fibroblasts and platelets to collagen. Crossblocking studies indicate that P1H5 and eBioY418 recognize different epitopes. Applications Reported: This P1H5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This P1H5 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirica...
CD49b (Integrin alpha 2, Integrin alpha 2 chain, VLA-2 alpha chain, HM alpha 2) is a member of the integrin family. It is a glycoprotein with molecular weight of 150 kD, and it complexes with CD29 (Integrin beta 1) to form the heterodimeric integrin VLA-2 (integrin alpha 2 beta 1, or GPIa/IIa) complex. VLA-2 is an extracellular receptor for laminin, collagen, and fibronectin, and interaction with its ligands results in the activation of intracellular signaling pathways. It has reported roles in VEGF-induced angiogenesis in vivo, as well as adhesion and lymphocyte activation. CD49b is expressed by NK cells, NK-T cells, monocytes, platelets, and epithelial cells. It is also expressed on adaptive immune cells such as T and B cells, specifically on a subset of CD4+ T cells in the spleen, on intraepithelial and lamina propria lymphocytes in the intestine, as well as on a population of peripheral CD4+ type 1 T regulatory (Tr1) cells that co-express LAG-3.Specifications
CD49b (Integrin alpha 2) | |
5 μL/Test | |
Flow Cytometry | |
PE | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P17301 | |
3673 | |
Primary | |
Affinity chromatography |
P1H5 | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
Itga2 | |
alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 | |
Itga2 | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.