Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD49b Mouse anti-Human, BV750, Clone: AK-7 (also known as AK7), BD Biosciences™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 747215
Description
CD49b Monoclonal antibody specifically detects CD49b in Human samples. It is validated for Flow Cytometry
Specifications
CD49b | |
Monoclonal | |
Brilliant Violet 750 | |
Integrin alpha-2; ITGA2; VLA-2 alpha; Platelet GPIa; Collagen receptor; BR | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
Flow Cytometry | |
AK-7 (also known as AK7) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
RUO | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction