Learn More
CD49b Mouse, anti-Mouse, R718, Clone: HMA2, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 751927
Description
The HMα2 antibody reacts with integrin α2 chain (CD49b), the 150-kDa transmembrane glycoprotein that non-covalently associates with the integrin β1 subunit (CD29) to form the integrin α2β1 complex known as VLA-2. VLA-2, a receptor for collagen and laminin, is expressed on some splenic CD4+ T lymphocytes and NK-T cells, intestinal intraepithelial and lamina propria lymphocytes, splenic NK cells, epithelial cells, and platelets| but it is not on thymocytes or Peyer's-patch or lymphnode lymphocytes. The expression of VLA-2 is upregulated on lymphocytes in response to mitogens. The HMα2 antibody has been reported to partially block the interaction of T-cell blasts, but not NK cells, with collagen. Purified HMα2 mAb blocks the staining of splenic NK cells by the anti-CD49b/Pan-NK Cells mAb DX5 (Cat. No. 553858, for the PE conjugate). Therefore, mAb HMα2 may be used like the DX5 mAb for identification of NK cells.

Specifications
CD49b | |
Monoclonal | |
R718 | |
50 μg | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
HMA2 | |
Mouse | |
RUO | |
Mouse | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.