Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49c (Integrin α3) Mouse anti-Human, BV480, Clone: P1B5 (also known as P1B5), BD OptiBuild™

Catalog No. BDB755287 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB755287 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB755287 Supplier BD Biosciences Supplier No. 755287
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CD49c (Integrin α3) Monoclonal antibody specifically detects CD49c (Integrin α3) in Human samples. It is validated for Flow cytometry (Qualified)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49c (Integrin α3)
Applications Flow Cytometry
Classification Monoclonal
Clone P1B5 (also known as P1B5)
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 480
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias ITGA3; integrin alpha 3; alpha 3 subunit of VLA-3 receptor; VLA3a
Host Species Mouse
Immunogen Human WI-38 VA13 or HT-1080 Cells
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3675
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.