Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD49c (Integrin α3) Mouse anti-Human, BV711, Clone: P1B5 (also known as P1B5), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 755291
This item is not returnable.
View return policy
Description
CD49c (Integrin α3) Monoclonal antibody specifically detects CD49c (Integrin α3) in Human samples. It is validated for Flow cytometry (Qualified)
Specifications
CD49c (Integrin α3) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
3675 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
P1B5 (also known as P1B5) | |
Brilliant Violet 711 | |
ITGA3; integrin alpha 3; alpha 3 subunit of VLA-3 receptor; VLA3a | |
Human WI-38 VA13 or HT-1080 Cells | |
50 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction