Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d (Integrin α4 chain) Rat anti-Mouse, BUV615, Clone: R1-2, BD OptiBuild™

Catalog No. BDB752352 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752352 1 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752352 Supplier BD Biosciences Supplier No. 752352
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD49d (Integrin α4 chain) Monoclonal antibody specifically detects CD49d (Integrin α4 chain) in Mouse samples. It is validated for Flow Cytometry.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49d (Integrin α4 chain)
Applications Flow Cytometry
Classification Monoclonal
Clone R1-2
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 615
Description Ms CD49d BUV615 R1-2 50 μg
Host Species Rat
Immunogen AKR/Cum mouse spontaneous T lymphoma line TK1
Quantity 1 x 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity CD49d (Integrin α4 chain)
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Isotype IgG2b κ
Reactivity Mouse
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.