Learn More
Invitrogen™ CD49d (Integrin alpha 4) Monoclonal Antibody (R1-2), PE, eBioscience™, Invitrogen™
Description
Description: The R1-2 monoclonal antibody reacts with mouse CD49d, the 150 kDa integrin alpha4 subunit. The complex of CD49d non-covalently associated with integrin beta1 (CD29), also known as VLA-4, is a receptor for fibronectin and VCAM-1 (CD106). This complex is expressed by thymocytes, peripheral lymphocytes, monocytes and eosinophils. CD49d also associates with integrin b7 and binds to the Mucosal vascular Addressin (MAdCAM-1). Applications Reported: The R1-2 antibody has been reported for use in flow cytometric analysis. Applications Tested: The R1-2 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD49d (Integrin alpha 4) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | R1-2 |
| Concentration | 0.2 mg/mL |
| Conjugate | PE |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | ITGA4 |
| Gene Accession No. | Q00651 |
| Gene Alias | 269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.