Learn More
CD49d Monoclonal Antibody (PS/2), Functional Grade, In Vivo GOLD™, Leinco Technologies™

Rat Monoclonal Antibody
Supplier: Leinco Technologies Inc C797100MG


Description
Specificity: PS/2 activity is directed against mouse CD49d and is cross reactive against human CD49d. Endotoxin Level: <1.0 EU/μg as determined by the LAL method. Purity: ≥95% monomer by analytical SEC and >95% by SDS Page. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -70C. Avoid Repeated Freeze Thaw Cycles.
CD49d Monoclonal antibody specifically detects CD49d in Human, Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunohistochemistry, ImmunoprecipitationSpecifications
CD49d | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
P13612, Q00651 | |
Rat | |
Protein A/G | |
RUO | |
16401, 3676 | |
4°C short term, -80°C long term | |
Liquid |
Flow Cytometry, Functional Assay, Immunohistochemistry, Immunoprecipitation | |
PS/2 | |
Functional Grade | |
ITGA4 | |
269C wild type; alpha 4 subunit of VLA-4 receptor; antigen CD49D, alpha-4 subunit of VLA-4 receptor; CD49 antigen-like family member D; CD49d; IA4; integrin alpha 4; integrin alpha-4; integrin alpha4B; Integrin alpha-IV; integrin subunit alpha 4; integrin, alpha 4; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); Itga4; Itga4B; ITGA-IV; LPAM subunit alpha; lymphocyte Peyer patch adhesion molecules subunit alpha; very late activation protein 4 receptor, alpha 4 subunit; VLA-4 receptor, alpha 4 subunit; VLA-4 subunit alpha | |
P815 DBA/2 murine mastocytoma cells. | |
100 mg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.