Learn More
CD49d Mouse anti-Baboon, Equine, Feline, Human, Brilliant Ultraviolet 737, Clone: 9F10, BD Horizon™
Shop All BD Biosciences ProductsDescription
The 9F10 monoclonal antibody specifically reacts with the integrin α 4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β 1 (CD29) or β 7. The α 4β 1 integrin (VLA-4) is expressed on lymphocytes, monocytes, thymocytes, NK cells, and several B- and T-cell lines, and mediates binding to VCAM-1 (CD106) and the CS-1 region of fibronectin. The α β 7 also binds fibronectin and VCAM-1, and has been shown in the mouse to preferentially bind the mucosal vascular addressin molecule, MAdCAM-1. This antibody is useful for studies of the expression by and function of cells that express α 4 chain-containing integrins. This clone cross-reacts with a subset of peripheral blood lymphocytes, monocytes, and some granulocytes of baboon and both rhesus and cynomolgus macaque monkeys. The distribution on leukocytes is similar to that observed with human peripheral blood leukocytes.
Specifications
Specifications
| Antigen | CD49d |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 9F10 |
| Conjugate | Brilliant Ultraviolet 737 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Regulatory Status | RUO |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.