Learn More
CD49d Mouse anti-Human, APC, Clone: 9F10, BD
Mouse Monoclonal Antibody
$164.70 - $390.00
Specifications
| Antigen | CD49d |
|---|---|
| Clone | 9F10 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | APC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
BDB561892
|
BD Biosciences
561892 |
25 Tests |
Each for $164.70
|
|
|||||
|
BDB559881
|
BD Biosciences
559881 |
100 Tests |
Each for $390.00
|
|
|||||
Description
The 9F10 monoclonal antibody specifically reacts with the integrin α4 chain, that is expressed as a heterodimer with either of two β integrin subunits, β1 (CD29) or β7. The α4β1 integrin (VLA-4) is expressed on lymphocytes, monocytes, thymocytes, NK cells, and several B- and T-cell lines, and mediates binding to VCAM-1 (CD106) and the CS-1 region of fibronectin. The αβ7 also binds fibronectin and VCAM-1, and has been shown in the mouse to preferentially bind the mucosal vascular addressin molecule, MAdCAM-1. This antibody is useful for studies of the expression by and function of cells that express α4 chain-containing integrins.
Specifications
| CD49d | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 9F10 | |
| Monoclonal | |
| Mouse | |
| Baboon, Bovine, Canine, Cynomolgus, Equine, Feline, Human, Rhesus Monkey, Sheep | |
| IgG1 κ | |
| Affinity Purified | |
| Integrin α4 chain; Integrin alpha 4; ITGA4; IA4; alpha 4 subunit of VLA-4 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.