Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d Mouse, anti-Human, R718, Clone: L25, BD OptiBuild™

Catalog No. BDB752156 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752156 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752156 Supplier BD Biosciences Supplier No. 752156
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The L25 monoclonal antibody specifically recognizes CD49d which is also known as Integrin alpha-4 (α4 Integrin) or the alpha chain of the Very-Late Antigen-4 (VLA-4α). CD49d is a ∼150 kDa type I transmembrane glycoprotein that is encoded by ITGA4 and belongs to the integrin family of cell adhesion molecules. VLA-4, like other integrins, is a noncovalently associated heterodimeric glycoprotein composed of α and β subunits and is involved in cell-cell and cell-extracellular matrix interactions. The β chain of the VLA-4 complex is the CD29 antigen, a 130 kDa glycoprotein. The CD29 antigen, also known as the β1 subunit, is common to the VLA family of integrins. When acting as a matrix receptor, the CD49d antigen binds to CS-1, an alternatively spliced domain of fibronectin. When functioning as a cell receptor, the CD49d antigen binds to the vascular cell-adhesion molecule-1 (VCAM-1). The interaction between the CD49d antigen and VCAM-1 is known to play an important role in stabilizing the adhesion of lymphocytes to endothelial cells and in mediating B-lymphocyte precursor/bone marrow stromal cell adhesion. The CD49d antigen, when associated with the β7 integrin, forms the α4/β7 integrin lymphocyte homing receptor for Peyer's patches, binding to the mucosal vascular addressin MAdCAM-1. The CD49d antigen is also involved in CD3-dependent CD4+ T-lymphocyte activation via its interaction with fibronectin. The CD49d antigen is primarily expressed on T and B lymphocytes and weakly expressed on monocytes. The L25 antibody can block or enhance fibronectin-stimulated T-lymphocyte proliferation. It immunoprecipitates three proteins of 150 kDa, 85 kDa, and 75 kDa under both reducing and nonreducing conditions from HPB-ALL cells, B lymphoblasts, peripheral blood lymphocytes, and IL-2-dependent cell lines.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone L25
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.