Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49d Mouse, anti-Rat, R718, Clone: MRA4-1, BD OptiBuild™

Catalog No. BDB752239 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752239 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752239 Supplier BD Biosciences Supplier No. 752239
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MRα4-1 antibody reacts with the integrin α4 chain, which is expressed as a heterodimer with either of two β chains, β1 or β7. The α4β1 integrin (VLA-4, CD49d/CD29) is expressed on peripheral T and B lymphocytes, thymocytes, and monocytes| while the α4β7 integrin (LPAM-1) is expressed on peripheral lymphocytes. These integrins mediate a variety of cell-cell and cell-matrix interactions, recognizing the ligands CD106 (VCAM-1), MAdCAM-1, and fibronectin. It has been reported that soluble mAb MRα4-1 partially inhibits in vitro binding of VCAM-1 and mast cells to fibronectin and inhibits the enhanced degranulation of IgE-sensitized RBL-2H3 basophilic leukemia cell line induced on fibronectin-coated plates. Furthermore, plate-bound MRα4-1 antibody enhances the degranulation of IgE-sensitized RBL-2H3 cells| and subcutaneous injection of MRα4-1 mAb, along with anti-CD49e and anti-CD61 antibodies, inhibits experimentally induced passive cutaneous anaphylaxis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49d
Applications Flow Cytometry
Classification Monoclonal
Clone MRA4-1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.