Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD49e Armenian Hamster anti-Mouse, BV711, Clone: HM alpha 5-1, BD OptiBuild™

Catalog No. BDB743113 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743113 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743113 Supplier BD Biosciences Supplier No. 743113
Add to Cart
Add to Cart

Armenian Hamster Monoclonal Antibody

CD49e Monoclonal antibody specifically detects CD49e in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e
Applications Flow Cytometry
Classification Monoclonal
Clone HM alpha 5-1
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 711
Host Species Armenian Hamster
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Refrigerate (2 to 8°C)
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.