Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD49e (Integrin alpha 5) Monoclonal Antibody (eBioSAM-1 (SAM-1, SAM1)), eBioscience™, Invitrogen™

Catalog No. 14049637
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-049-637 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-049-637 Supplier Invitrogen™ Supplier No. 14049637
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The eBioSAM-1 MAb reacts with human integrin alpha 5, also known as fibronectin receptor alpha chain, very late activation antigen 5 alpha, and CD49e. Integrins are composed of an alpha chain and a beta chain, which non-covalently associate to form the functional integrin. Integrin heterodimers participate in cell surface-mediated signaling and adhesion functions. Integrin alpha 5 undergoes post-translational cleavage in its extracellular domain to yield disulfide linked light and heavy chains that join with Integrin beta 1 (CD29) to form the fibronectin receptor, also known as the very late activation antigen-5 (VLA-5) complex. Integrin alpha 5 is expressed on thymocytes, T cells, monocytes, platelets, early B cells, and activated B cells.

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD49e (Integrin alpha 5)
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone eBioSAM-1 (SAM-1, SAM1)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Itga5
Gene Accession No. P08648
Gene Alias 2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A
Gene Symbols Itga5
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3678
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.